Gene Transcripts

Gene Symbol
Ostn
Entrez Gene ID
239790
Gene Name
osteocrin
Gene Aliases
Ostc
Chromosome Location
Chr. 16: 27306983 - 27351209 on Build GRCm38
UniGene ID
Mm.390880
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198112.2 NP_932780.1 4 409
GenBank mRNA AY395730.1 - 4 409
AY573932.1 - 4 561
BC117063.1 - 4 397 40126382
BC117089.1 - 4 397 40126408

View Full Product Details