Gene Transcripts

Gene Symbol
Tcl1b4
Entrez Gene ID
27380
Gene Name
T cell leukemia/lymphoma 1B, 4
Gene Aliases
AA675342
Chromosome Location
Chr. 12: 105201957 - 105206993 on Build GRCm38
UniGene ID
Mm.103652
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013774.2 NP_038802.1 3 417
GenBank mRNA AF195492.1 - 3 417

View Full Product Details