Gene Transcripts

Gene Symbol
Gimap9
Entrez Gene ID
317758
Gene Name
GTPase, IMAP family member 9
Gene Aliases
A630002K24, BB145400
Chromosome Location
Chr. 6: 48676121 - 48679116 on Build GRCm38
UniGene ID
Mm.116971
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174960.2 NP_777620.1 2 824
GenBank mRNA AK041336.1 - 2 813
BC038043.1 - 2 746
BC096680.1 - 2 808 1345477

View Full Product Details