Gene Transcripts

Gene Symbol
Lrch1
Entrez Gene ID
380916
Gene Name
leucine-rich repeats and calponin homology (CH) domain containing 1
Gene Aliases
4832412D13Rik, AI552548, AW215503, Chdc1, mKIAA1016
Chromosome Location
Chr. 14: 74754673 - 74948039 on Build GRCm38
UniGene ID
Mm.288868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033439.3 NP_001028611.2 3 748
NM_001252132.1 NP_001239061.1 3 748
GenBank mRNA AK129264.1 - 3 737
AK136026.1 - 3 354
AK143909.1 - 3 747
AK161894.1 - 3 547

View Full Product Details