Gene Transcripts
- Gene Symbol
- LYPLA1
- Entrez Gene ID
- 10434
- Gene Name
- lysophospholipase I
- Gene Aliases
- APT-1, APT1, LPL-I, LPL1, hAPT1
- Chromosome Location
- Chr. 8: 54046367 - 54102017 on Build GRCh38
- UniGene ID
- Hs.744046
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001279356.1 |
NP_001266285.1
|
7, 8 |
724 |
|
NM_001279357.1 |
NP_001266286.1
|
7, 8 |
778 |
|
NM_001279358.1 |
NP_001266287.1
|
8, 9 |
784 |
|
NM_001279359.1 |
NP_001266288.1
|
8, 9 |
786 |
|
NM_001279360.1 |
NP_001266289.1
|
8, 9 |
768 |
|
NM_006330.3 |
NP_006321.1
|
8, 9 |
826 |
|
XM_005251127.4 |
XP_005251184.1
|
8, 9 |
626 |
|
XM_017012956.1 |
XP_016868445.1
|
7, 8 |
737 |
|
XM_017012957.1 |
XP_016868446.1
|
9, 10 |
776 |
|
XM_017012958.1 |
XP_016868447.1
|
8, 9 |
661 |
|
XM_017012959.1 |
XP_016868448.1
|
9, 10 |
825 |
|
XM_017012960.1 |
XP_016868449.1
|
7, 8 |
643 |
|
GenBank mRNA |
AF035293.1 |
- |
2, 3 |
255 |
|
AF052112.1 |
- |
8, 9 |
826 |
|
AF077198.1 |
- |
8, 9 |
637 |
|
AF077199.1 |
- |
7, 8 |
589 |
|
AF081281.1 |
- |
8, 9 |
667 |
|
AF291053.1 |
- |
8, 9 |
650 |
|
AK289400.1 |
- |
8, 9 |
710 |
|
AK298206.1 |
- |
7, 8 |
568 |
|
AK313739.1 |
- |
8, 9 |
663 |
|
BC008652.2 |
- |
8, 9 |
714 |
3865775 |
BC010397.1 |
- |
8, 9 |
638 |
4109335 |
CR457103.1 |
- |
8, 9 |
632 |
|
View Full Product Details