Gene Transcripts
- Gene Symbol
- Fbxw27
- Entrez Gene ID
- 76998
- Gene Name
- F-box and WD-40 domain protein 27
- Gene Aliases
- 1700124P09Rik
- Chromosome Location
- Chr. 9: 109765655 - 109790653 on Build GRCm38
- UniGene ID
- Mm.442009
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CJ082096.1 |
- |
4 |
362 |
|
CO804891.1 |
- |
1 |
39 |
|
DV663185.1 |
- |
4 |
342 |
|
View Full Product Details