Gene Transcripts

Gene Symbol
FARS2
Entrez Gene ID
10667
Gene Name
phenylalanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD14, FARS1, HSPC320, PheRS
Chromosome Location
Chr. 6: 5261001 - 5771592 on Build GRCh38
UniGene ID
Hs.484547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318872.1 NP_001305801.1 4 1163
NM_006567.4 NP_006558.1 4 1157
XM_005248812.3 XP_005248869.1 5 1221
XM_006714966.2 XP_006715029.1 4 1132
XM_011514247.2 XP_011512549.1 4 1132
XM_011514248.2 XP_011512550.1 4 1132
XM_011514249.1 XP_011512551.1 4 1132
XM_011514251.2 XP_011512553.1 4 1132
XM_017010186.1 XP_016865675.1 4 1940
XM_017010187.1 XP_016865676.1 5 1942
GenBank mRNA AF097441.1 - 3 813
AF161438.1 - 3 482
AK223423.1 - 4 957
AK312454.1 - 4 1137
BC020239.1 - 4 957 5258249
BC021112.1 - 4 957 5088776
CR542279.1 - 3 801

View Full Product Details