Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006662.2 | NP_006653.2 | 12 | 2117 | |
GenBank mRNA | AB621816.1 | - | 12 | 2184 | |
AF143946.1 | - | 8 | 1351 | ||
AK128030.1 | - | 12 | 2117 | ||
BC025284.1 | - | 9 | 1594 | ||
BC063578.1 | - | 10 | 1911 |