Gene Transcripts

Gene Symbol
MORF4L1
Entrez Gene ID
10933
Gene Name
mortality factor 4 like 1
Gene Aliases
Eaf3, FWP006, HsT17725, MEAF3, MORFRG15, MRG15, S863-6
Chromosome Location
Chr. 15: 78872781 - 78897739 on Build GRCh38
UniGene ID
Hs.374503
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001265604.1 NP_001252533.1 3 317
NM_001265605.1 NP_001252534.1 3 217
NM_006791.3 NP_006782.1 3 364
NM_206839.2 NP_996670.1 3 364
XM_017021872.1 XP_016877361.1 3 329
GenBank mRNA AF070664.1 - 3 171
AF100615.1 - 3 258
AF109188.1 - 3 153
AF161546.1 - 3 150
AF167173.1 - 3 166
AF218011.1 - 3 269
AK098485.1 - 3 284
AK296650.1 - 3 255
AK300789.1 - 3 301
AK310181.1 - 2 102
AK310592.1 - 3 163
AK310702.1 - 3 172
AL137697.1 - 3 158
AY148481.1 - 3 258
BC002936.2 - 3 245 3955120
BC022845.2 - 3 245 2988803
BC067826.1 - 3 140 4824022
BC141840.1 - 3 172 8860303
BG771854.1 - 3 374
BX382045.2 - 3 161
DB455429.1 - 3 217

View Full Product Details