Gene Transcripts
- Gene Symbol
- LILRA1
- Entrez Gene ID
- 11024
- Gene Name
- leukocyte immunoglobulin like receptor A1
- Gene Aliases
- CD85I, LIR-6, LIR6
- Chromosome Location
- Chr. 19: 54593576 - 54602381 on Build GRCh38
- UniGene ID
- Hs.710507
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF025529.1 |
- |
8 |
1249 |
|
AF025530.1 |
- |
10 |
1861 |
|
AK290919.1 |
- |
10 |
1877 |
|
View Full Product Details