Gene Transcripts

Gene Symbol
TREH
Entrez Gene ID
11181
Gene Name
trehalase
Gene Aliases
TRE, TREA
Chromosome Location
Chr. 11: 118658232 - 118679672 on Build GRCh38
UniGene ID
Hs.129712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301065.1 NP_001287994.1 7 768
NM_007180.2 NP_009111.2 8 861
GenBank mRNA AB000824.1 - 8 871
AK091981.1 - 6 730
AK223140.1 - 8 835
AK223143.1 - 8 839
AK309565.1 - 8 835
BC109206.2 - 7 762 40006501

View Full Product Details