Gene Transcripts

Gene Symbol
Rhoc
Entrez Gene ID
295342
Gene Name
ras homolog family member C
Gene Aliases
Chromosome Location
Chr. 2: 207271692 - 207278010 on Build Rnor_6.0
UniGene ID
Rn.162614
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106461.1 NP_001099931.1 4 641
XM_006233074.1 XP_006233136.1 4 615
GenBank mRNA BC166849.1 - 4 657

View Full Product Details