Gene Transcripts

Gene Symbol
Rsbn1l
Entrez Gene ID
311987
Gene Name
round spermatid basic protein 1-like
Gene Aliases
RGD1308748
Chromosome Location
Chr. 4: 10748745 - 10809142 on Build Rnor_6.0
UniGene ID
Rn.139850
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135872.1 NP_001129344.1 3 1421

View Full Product Details