Gene Transcripts

Gene Symbol
TMEM258
Entrez Gene ID
746
Gene Name
transmembrane protein 258
Gene Aliases
C11orf10
Chromosome Location
Chr. 11: 61789130 - 61792613 on Build GRCh38
UniGene ID
Hs.437779
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014206.3 NP_055021.1 2 121
GenBank mRNA AF070661.1 - 2 121
AF085986.1 - 2 94
AF086763.1 - 2 117
AK291683.1 - 2 118
BC002750.2 - 2 92 3633279
BC015968.1 - 2 122 4073907

View Full Product Details