Gene Transcripts

Gene Symbol
CHIT1
Entrez Gene ID
1118
Gene Name
chitinase 1
Gene Aliases
CHI3, CHIT, CHITD
Chromosome Location
Chr. 1: 203216079 - 203229732 on Build GRCh38
UniGene ID
Hs.201688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256125.1 NP_001243054.2 8 991
NM_001270509.1 NP_001257438.1 9 1000
NM_003465.2 NP_003456.1 9 1048
NR_045784.1 9 1048
NR_045785.1 9 1048
GenBank mRNA AK055165.1 - 8 931
AK131093.1 - 10 5183
AK225245.1 - 9 940
AK309341.1 - 9 990
AK314331.1 - 9 987
BC069614.1 - 8 917
BC103695.1 - 9 964 40005693
BC105680.1 - 9 964 40005699
BC105681.1 - 9 964 40005697
BC105682.1 - 9 964 40005696
BP297140.1 - 4 386
U29615.1 - 9 964
U62662.1 - 9 961

View Full Product Details