Gene Transcripts

Gene Symbol
CYP4V2
Entrez Gene ID
285440
Gene Name
cytochrome P450 family 4 subfamily V member 2
Gene Aliases
BCD, CYP4AH1
Chromosome Location
Chr. 4: 186191520 - 186213463 on Build GRCh38
UniGene ID
Hs.587231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207352.3 NP_997235.3 4 817
XM_005262935.3 XP_005262992.1 4 602
XM_017008037.1 XP_016863526.1 3 575
GenBank mRNA AK122600.1 - 4 552
AK126473.1 - 5 486
AY422002.1 - 4 817
BC060857.1 - 4 683 30333559
BX648730.1 - 1 321

View Full Product Details