Gene Transcripts

Gene Symbol
NRGN
Entrez Gene ID
4900
Gene Name
neurogranin
Gene Aliases
RC3, hng
Chromosome Location
Chr. 11: 124739933 - 124747206 on Build GRCh38
UniGene ID
Hs.524116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126181.1 NP_001119653.1 4 940
NM_006176.2 NP_006167.1 4 937
GenBank mRNA BC002835.2 - 4 828
Y09689.1 - 4 914

View Full Product Details