Gene Transcripts
- Gene Symbol
- ZMAT5
- Entrez Gene ID
- 55954
- Gene Name
- zinc finger matrin-type 5
- Gene Aliases
- SNRNP20, U11/U12-20K, ZC3H19
- Chromosome Location
- Chr. 22: 29730956 - 29767011 on Build GRCh38
- UniGene ID
- Hs.713647
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003692.1 |
NP_001003692.1
|
3, 4 |
320 |
|
NM_001318129.1 |
NP_001305058.1
|
3, 4 |
274 |
|
NM_019103.2 |
NP_061976.1
|
4, 5 |
435 |
|
GenBank mRNA |
AA312132.1 |
- |
3, 4 |
320 |
|
AK292671.1 |
- |
3, 4 |
301 |
|
AL160311.1 |
- |
3, 4 |
319 |
|
BC009717.1 |
- |
3, 4 |
285 |
3890730 |
BG285784.1 |
- |
4, 5 |
399 |
|
BQ931527.1 |
- |
3, 4 |
248 |
|
BU598307.1 |
- |
3, 4 |
322 |
|
CR456353.1 |
- |
2, 3 |
207 |
|
CR457393.1 |
- |
2, 3 |
184 |
|
DT217179.1 |
- |
4, 5 |
451 |
|
View Full Product Details