Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000021.3 | NP_000012.1 | 6 | 797 | |
NM_007318.2 | NP_015557.2 | 6 | 785 | ||
XM_005267864.2 | XP_005267921.1 | 6 | 791 | ||
XM_005267866.1 | XP_005267923.1 | 6 | 780 | ||
XM_011536971.2 | XP_011535273.1 | 6 | 689 | ||
XM_011536972.2 | XP_011535274.1 | 6 | 1053 | ||
XM_011536973.1 | XP_011535275.1 | 6 | 1041 | ||
XM_011536974.1 | XP_011535276.1 | 6 | 682 | ||
GenBank mRNA | AF416717.1 | - | 4 | 513 | |
AK122722.1 | - | 4 | 2066 | ||
AK223173.1 | - | 4 | 527 | ||
AK312531.1 | - | 6 | 781 | ||
BC011729.2 | - | 6 | 766 | 4423098 | |
KU178285.1 | - | 4 | 501 | ||
KU178286.1 | - | 4 | 513 | ||
L42110.1 | - | 6 | 761 | ||
L76517.1 | - | 6 | 1054 | ||
U40379.1 | - | 4 | 501 | ||
U40380.1 | - | 4 | 501 |