Gene Transcripts

Gene Symbol
NOM1
Entrez Gene ID
64434
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
C7orf3, PPP1R113, SGD1
Chromosome Location
Chr. 7: 156949723 - 156973182 on Build GRCh38
UniGene ID
Hs.15825
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138400.1 NP_612409.1 8 2165
GenBank mRNA AY576779.1 - 8 2165
BC007902.2 - 4 456

View Full Product Details