Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003345.4 | NP_003336.1 | 4 | 358 | |
NM_194259.2 | NP_919235.1 | 5 | 615 | ||
NM_194260.2 | NP_919236.1 | 4 | 478 | ||
NM_194261.2 | NP_919237.1 | 4 | 330 | ||
XM_005255540.2 | XP_005255597.1 | 4 | 349 | ||
XM_005255541.4 | XP_005255598.1 | 4 | 755 | ||
XM_011522644.2 | XP_011520946.1 | 5 | 595 | ||
XM_011522645.1 | XP_011520947.1 | 4 | 460 | ||
XM_017023640.1 | XP_016879129.1 | 4 | 755 | ||
GenBank mRNA | AB208988.1 | - | 4 | 439 | |
AJ002385.1 | - | 4 | 278 | ||
AK093616.1 | - | 4 | 289 | ||
AK291118.1 | - | 4 | 349 | ||
AL833737.1 | - | 1 | 1155 | ||
BC000427.2 | - | 4 | 262 | 2819764 | |
BC004429.1 | - | 4 | 262 | 2819764 | |
BC051289.1 | - | 4 | 292 | ||
BE262845.1 | - | 5 | 597 | ||
BI523501.1 | - | 4 | 468 | ||
BT006932.1 | - | 3 | 200 | ||
BT007010.1 | - | 3 | 200 | ||
BU543455.1 | - | 4 | 350 | ||
CD249028.1 | - | 4 | 299 | ||
CX788607.1 | - | 4 | 434 | ||
D45050.1 | - | 4 | 317 | ||
U29092.1 | - | 3 | 224 | ||
U31882.1 | - | 4 | 293 | ||
U31933.1 | - | 4 | 318 | ||
U38785.1 | - | 4 | 292 | ||
U45328.1 | - | 4 | 323 | ||
U66818.1 | - | 4 | 287 | ||
U66867.1 | - | 4 | 1006 | ||
X96427.1 | - | 4 | 323 |