Gene Transcripts

Gene Symbol
ADAT3
Entrez Gene ID
113179
Gene Name
adenosine deaminase, tRNA specific 3
Gene Aliases
FWP005, MRT36, MST121, MSTP121, S863-5, TAD3
Chromosome Location
Chr. 19: 1905372 - 1913447 on Build GRCh38
UniGene ID
Hs.144980
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138422.2 NP_612431.2 2 1492
GenBank mRNA BC011824.1 - 2 1446

View Full Product Details