Gene Transcripts

Gene Symbol
POU5F2
Entrez Gene ID
134187
Gene Name
POU domain class 5, transcription factor 2
Gene Aliases
SPRM-1
Chromosome Location
Chr. 5: 93740309 - 93741603 on Build GRCh38
UniGene ID
Hs.678995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153216.1 NP_694948.1 1 697
GenBank mRNA AK098546.1 - 1 697
BC029532.1 - 1 657 5295125
BX641019.1 - 1 217
Z21965.1 - 1 157

View Full Product Details