Gene Transcripts

Gene Symbol
TIPRL
Entrez Gene ID
261726
Gene Name
TOR signaling pathway regulator
Gene Aliases
TIP, TIP41, TIPRL1
Chromosome Location
Chr. 1: 168178845 - 168202114 on Build GRCh38
UniGene ID
Hs.209431
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031800.2 NP_001026970.1 4 641
NM_152902.4 NP_690866.1 4 641
GenBank mRNA AB097034.1 - 4 524
AK313520.1 - 4 641
AL049670.1 - 4 553
BC009506.2 - 4 431 3532344

View Full Product Details