Gene Transcripts

Gene Symbol
SIMC1
Entrez Gene ID
375484
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25, OOMA1, PLEIAD
Chromosome Location
Chr. 5: 176238359 - 176345991 on Build GRCh38
UniGene ID
Hs.719847
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC037298.1 - 4, 5 1798
KF151170.1 - 2, 3 1457

View Full Product Details