Gene Transcripts

Gene Symbol
INSC
Entrez Gene ID
387755
Gene Name
inscuteable homolog (Drosophila)
Gene Aliases
Chromosome Location
Chr. 11: 15111416 - 15269675 on Build GRCh38
UniGene ID
Hs.591997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031853.4 NP_001027024.3 7 901
NM_001042536.2 NP_001036001.1 7 825
NM_001278313.1 NP_001265242.1 6 787
NM_001278314.1 NP_001265243.1 7 892
NM_001278315.1 NP_001265244.1 6 787
XM_006718227.3 XP_006718290.1 8 954
XM_011520085.1 XP_011518387.1 7 829
XM_011520086.1 XP_011518388.1 7 829
XM_017017697.1 XP_016873186.1 8 866
XM_017017698.1 XP_016873187.1 7 829
GenBank mRNA AB231745.1 - 6 781
AB231746.1 - 7 886
AB236158.1 - 7 825
AB236159.1 - 7 901
BC127700.1 - 6 787 40127962

View Full Product Details