Gene Transcripts

Gene Symbol
CD99P1
Entrez Gene ID
401577
Gene Name
CD99 molecule pseudogene 1
Gene Aliases
CD99L1, CXYorf12, MIC2R, NCRNA00103
Chromosome Location
Chr. Y: 2609265 - 2657229 on Build GRCh38
UniGene ID
Hs.444541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033381.1 10 1099
GenBank mRNA AK023533.1 - 6 2737
BC019893.1 - 10 970

View Full Product Details