Gene Transcripts

Gene Symbol
SCGB1D4
Entrez Gene ID
404552
Gene Name
secretoglobin family 1D member 4
Gene Aliases
Chromosome Location
Chr. 11: 62296282 - 62299064 on Build GRCh38
UniGene ID
Hs.473247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206998.1 NP_996881.1 3 360
GenBank mRNA AY236538.1 - 3 327
AY359052.1 - 3 360

View Full Product Details