Gene Transcripts
- Gene Symbol
- LOC100130394
- Entrez Gene ID
- 100130394
- Gene Name
- translation machinery associated 7 homolog (S. cerevisiae) pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 172656359 - 172656734 on Build GRCh38
- UniGene ID
- Hs.585199
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF275799.1 |
- |
1 |
97 |
|
- Gene Symbol
- TMA7
- Entrez Gene ID
- 51372
- Gene Name
- translation machinery associated 7 homolog
- Gene Aliases
- CCDC72, HSPC016
- Chromosome Location
- Chr. 3: 48440276 - 48444129 on Build GRCh38
- UniGene ID
- Hs.356440
- Species
- Homo sapiens
View Full Product Details