Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024574.3 | NP_078850.3 | 4 | 1814 | |
GenBank mRNA | AK026844.1 | - | 1 | 900 | |
AK289615.1 | - | 4 | 1722 | ||
AY358821.1 | - | 4 | 1386 | ||
BC019351.1 | - | 4 | 1710 | 3928768 |