Gene Transcripts

Gene Symbol
KRTAP1-1
Entrez Gene ID
81851
Gene Name
keratin associated protein 1-1
Gene Aliases
HB2A, KAP1.1, KAP1.1A, KAP1.1B, KAP1.6, KAP1.7, KRTAP1.1, KRTAP1A, hKAP1.7
Chromosome Location
Chr. 17: 41040559 - 41041461 on Build GRCh38
UniGene ID
Hs.247934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030967.2 NP_112229.1 1 815
GenBank mRNA AB052868.1 - 2 677
AB052934.1 - 1 815
AB055057.1 - 2 490
AJ406926.1 - 1 784

View Full Product Details