Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114107.4 | NP_001107579.1 | 2 | 327 | |
NM_001257962.1 | NP_001244891.1 | 2 | 327 | ||
NM_014476.5 | NP_055291.2 | 2 | 327 | ||
NR_047562.1 | 2 | 327 | |||
GenBank mRNA | AF002280.1 | - | 2 | 249 | |
AF002282.1 | - | 2 | 249 | ||
AF039018.1 | - | 2 | 273 | ||
AK129863.1 | - | 2 | 196 | ||
AK222966.1 | - | 2 | 231 | ||
AK313253.1 | - | 2 | 276 | ||
BC027870.1 | - | 2 | 254 | 5205215 | |
BI913867.1 | - | 2 | 327 | ||
BT007341.1 | - | 2 | 203 | ||
C03422.1 | - | 2 | 153 |