Gene Transcripts

Gene Symbol
CPPED1
Entrez Gene ID
55313
Gene Name
calcineurin like phosphoesterase domain containing 1
Gene Aliases
CSTP1
Chromosome Location
Chr. 16: 12659799 - 12803887 on Build GRCh38
UniGene ID
Hs.460002
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099455.1 NP_001092925.1 3 1019
NM_018340.2 NP_060810.2 4 1445
GenBank mRNA AK002013.1 - 4 1428
AK022710.1 - 4 1395
AK093854.1 - 4 1191
AK225617.1 - 4 1440
AK298662.1 - 3 849
BC006289.2 - 4 1429 3959701
BX640805.1 - 3 985

View Full Product Details