Gene Transcripts

Gene Symbol
CDC7
Entrez Gene ID
8317
Gene Name
cell division cycle 7
Gene Aliases
CDC7L1, HsCDC7, Hsk1, huCDC7
Chromosome Location
Chr. 1: 91500814 - 91525764 on Build GRCh38
UniGene ID
Hs.533573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134419.1 NP_001127891.1 12 1558
NM_001134420.1 NP_001127892.1 12 1686
NM_003503.3 NP_003494.1 12 1592
XM_005271241.2 XP_005271298.1 12 1554
XM_005271244.2 XP_005271301.1 11 1496
XM_017002425.1 XP_016857914.1 11 1458
XM_017002426.1 XP_016857915.1 11 1441
XM_017002427.1 XP_016857916.1 11 1400
GenBank mRNA AB003698.1 - 12 1559
AB209337.1 - 9 3686
AF005209.1 - 11 1427
AF015592.1 - 12 1572
AK298963.1 - 11 1504
AK312726.1 - 12 1648
AY312583.1 - 3 247
BC110526.1 - 12 1515 40034178
BC110527.1 - 12 1549 40034179
BC111044.1 - 12 1572 5518616

View Full Product Details