Gene Transcripts

Gene Symbol
PRR32
Entrez Gene ID
100130613
Gene Name
proline rich 32
Gene Aliases
CXorf64
Chromosome Location
Chr. X: 126819764 - 126821786 on Build GRCh38
UniGene ID
Hs.130370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122716.1 NP_001116188.1 2 129
GenBank mRNA BG200532.1 - 1 29
R10182.1 - 129

View Full Product Details