Gene Transcripts

Gene Symbol
NUTM2F
Entrez Gene ID
54754
Gene Name
NUT family member 2F
Gene Aliases
FAM22F, NUTMF
Chromosome Location
Chr. 9: 94318196 - 94328644 on Build GRCh38
UniGene ID
Hs.648183
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017561.1 NP_060031.1 7 1584
GenBank mRNA AL133071.1 - 6 1137
BC130390.1 - 7 1584 40146421
BC143798.1 - 7 1584 9052312

Gene Symbol
NUTM2G
Entrez Gene ID
441457
Gene Name
NUT family member 2G
Gene Aliases
FAM22G, NUTMG
Chromosome Location
Chr. 9: 96929004 - 96942290 on Build GRCh38
UniGene ID
Hs.652383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170741.1 NP_001164212.1 7 1539

View Full Product Details