Gene Transcripts

Gene Symbol
PTPMT1
Entrez Gene ID
114971
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Gene Aliases
DUSP23, MOSP, PLIP, PNAS-129
Chromosome Location
Chr. 11: 47565430 - 47573461 on Build GRCh38
UniGene ID
Hs.656205
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143984.1 NP_001137456.1 2 642
NM_175732.2 NP_783859.1 4 725
GenBank mRNA AF277187.1 - 2 139
AK294640.1 - 3 364
AY333987.1 - 3 340
BC014048.1 - 3 371
BC020242.1 - 4 554 4591438
BC107573.1 - 2 477
BQ050653.1 - 2 118
DR004030.1 - 4 583

View Full Product Details