Gene Transcripts

Gene Symbol
KRTAP10-7
Entrez Gene ID
386675
Gene Name
keratin associated protein 10-7
Gene Aliases
KAP10.7, KAP18.7, KRTAP18-7
Chromosome Location
Chr. 21: 44600597 - 44602174 on Build GRCh38
UniGene ID
Hs.473999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198689.2 NP_941962.1 1 1550
GenBank mRNA AJ566385.1 - 1 576

View Full Product Details