Gene Transcripts
- Gene Symbol
- GAGE12H
- Entrez Gene ID
- 729442
- Gene Name
- G antigen 12H
- Gene Aliases
- Chromosome Location
- Chr. X: 49579949 - 49587304 on Build GRCh38
- UniGene ID
- Hs.632813
- Species
- Homo sapiens
- Gene Symbol
- GAGE1
- Entrez Gene ID
- 2543
- Gene Name
- G antigen 1
- Gene Aliases
- CT4.1, GAGE-1
- Chromosome Location
- Chr. X: 49598992 - 49608536 on Build GRCh38
- UniGene ID
- Hs.278606
- Species
- Homo sapiens
- Gene Symbol
- GAGE12B
- Entrez Gene ID
- 729428
- Gene Name
- G antigen 12B
- Gene Aliases
- GAGE-12B
- Chromosome Location
- Chr. X: 49341183 - 49529921 on Build GRCh38
- UniGene ID
- Hs.632812
- Species
- Homo sapiens
- Gene Symbol
- GAGE12C
- Entrez Gene ID
- 729422
- Gene Name
- G antigen 12C
- Gene Aliases
- GAGE-12B
- Chromosome Location
- Chr. X: 49532177 - 49539541 on Build GRCh38
- UniGene ID
- Hs.632812
- Species
- Homo sapiens
- Gene Symbol
- GAGE12D
- Entrez Gene ID
- 100132399
- Gene Name
- G antigen 12D
- Gene Aliases
- GAGE-12B
- Chromosome Location
- Chr. X: 49541722 - 49549107 on Build GRCh38
- UniGene ID
- Hs.645554
- Species
- Homo sapiens
- Gene Symbol
- GAGE12E
- Entrez Gene ID
- 729431
- Gene Name
- G antigen 12E
- Gene Aliases
- GAGE-12B
- Chromosome Location
- Chr. X: 49551289 - 49558649 on Build GRCh38
- UniGene ID
- Hs.632812
- Species
- Homo sapiens
- Gene Symbol
- GAGE12F
- Entrez Gene ID
- 100008586
- Gene Name
- G antigen 12F
- Gene Aliases
- Chromosome Location
- Chr. X: 49560808 - 49568208 on Build GRCh38
- UniGene ID
- Hs.460641
- Species
- Homo sapiens
- Gene Symbol
- GAGE12G
- Entrez Gene ID
- 645073
- Gene Name
- G antigen 12G
- Gene Aliases
- AL4, CT4.7, GAGE-12G
- Chromosome Location
- Chr. X: 49570400 - 49577757 on Build GRCh38
- UniGene ID
- Hs.460641
- Species
- Homo sapiens
- Gene Symbol
- GAGE12I
- Entrez Gene ID
- 26748
- Gene Name
- G antigen 12I
- Gene Aliases
- AL4, CT4.7, GAGE-7B, GAGE7B
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.460641
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001477.1 |
NP_001468.1
|
4 |
323 |
|
- Gene Symbol
- GAGE12J
- Entrez Gene ID
- 729396
- Gene Name
- G antigen 12J
- Gene Aliases
- GAGE11
- Chromosome Location
- Chr. X: 49322030 - 49329387 on Build GRCh38
- UniGene ID
- Hs.645554
- Species
- Homo sapiens
- Gene Symbol
- GAGE13
- Entrez Gene ID
- 645051
- Gene Name
- G antigen 13
- Gene Aliases
- GAGE-12A, GAGE-13, GAGE12A
- Chromosome Location
- Chr. X: 49331603 - 49338952 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- GAGE2A
- Entrez Gene ID
- 729447
- Gene Name
- G antigen 2A
- Gene Aliases
- CT4.2, GAGE-2, GAGE-2A, GAGE2
- Chromosome Location
- Chr. X: 49589515 - 49596827 on Build GRCh38
- UniGene ID
- Hs.460641
- Species
- Homo sapiens
- Gene Symbol
- GAGE2B
- Entrez Gene ID
- 645037
- Gene Name
- G antigen 2B
- Gene Aliases
- CT4.2, GAGE-2, GAGE-2B
- Chromosome Location
- Chr. X: 49331615 - 49338933 on Build GRCh38
- UniGene ID
- Hs.714681
- Species
- Homo sapiens
- Gene Symbol
- GAGE2C
- Entrez Gene ID
- 2574
- Gene Name
- G antigen 2C
- Gene Aliases
- CT4.2, GAGE-2, GAGE-2C, GAGE2
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.460641
- Species
- Homo sapiens
- Gene Symbol
- GAGE2D
- Entrez Gene ID
- 729408
- Gene Name
- G antigen 2D
- Gene Aliases
- CT4.8, GAGE-2D, GAGE-8, GAGE8
- Chromosome Location
- Chr. X: 49560844 - 49568208 on Build GRCh38
- UniGene ID
- Hs.695912
- Species
- Homo sapiens
- Gene Symbol
- GAGE2E
- Entrez Gene ID
- 26749
- Gene Name
- G antigen 2E
- Gene Aliases
- GAGE-2E, GAGE8
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.695912
- Species
- Homo sapiens
- Gene Symbol
- GAGE3
- Entrez Gene ID
- 2575
- Gene Name
- G antigen 3
- Gene Aliases
- CT4.3
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U19144.1 |
- |
|
371 |
|
- Gene Symbol
- GAGE4
- Entrez Gene ID
- 2576
- Gene Name
- G antigen 4
- Gene Aliases
- CT4.4
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.460641
- Species
- Homo sapiens
- Gene Symbol
- GAGE5
- Entrez Gene ID
- 2577
- Gene Name
- G antigen 5
- Gene Aliases
- CT4.5
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.460641
- Species
- Homo sapiens
- Gene Symbol
- GAGE6
- Entrez Gene ID
- 2578
- Gene Name
- G antigen 6
- Gene Aliases
- CT4.6
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.632808
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001476.1 |
NP_001467.1
|
4 |
350 |
|
GenBank mRNA |
U19147.1 |
- |
4 |
350 |
|
- Gene Symbol
- GAGE7
- Entrez Gene ID
- 2579
- Gene Name
- G antigen 7
- Gene Aliases
- AL4, CT4.7, GAGE-7
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.460641
- Species
- Homo sapiens
- Gene Symbol
- GAGE8
- Entrez Gene ID
- 100101629
- Gene Name
- G antigen 8
- Gene Aliases
- CT4.8, GAGE-8
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.695912
- Species
- Homo sapiens
View Full Product Details