Gene Transcripts

Gene Symbol
LCNL1
Entrez Gene ID
401562
Gene Name
lipocalin like 1
Gene Aliases
Chromosome Location
Chr. 9: 136982993 - 136985758 on Build GRCh38
UniGene ID
Hs.695210
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207510.3 NP_997393.3 3 1465
GenBank mRNA AK097466.1 - 2 914
AK127160.1 - 3 1465
AK127236.1 - 2 3335

View Full Product Details