Gene Transcripts

Gene Symbol
DOCK10
Entrez Gene ID
55619
Gene Name
dedicator of cytokinesis 10
Gene Aliases
DRIP2, Nbla10300, ZIZ3
Chromosome Location
Chr. 2: 224765090 - 225042689 on Build GRCh38
UniGene ID
Hs.46578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290263.1 NP_001277192.1 44 4977
NM_014689.2 NP_055504.2 44 5120
XM_005246673.4 XP_005246730.1 44 4979
XM_006712617.3 XP_006712680.1 44 5144
XM_017004441.1 XP_016859930.1 44 5147
XM_017004442.1 XP_016859931.1 44 5123
XM_017004443.1 XP_016859932.1 44 5196
XM_017004444.1 XP_016859933.1 43 5103
XM_017004445.1 XP_016859934.1 44 4977
XM_017004446.1 XP_016859935.1 43 5001
XM_017004447.1 XP_016859936.1 44 5119
GenBank mRNA AF250396.1 - 4 489
AK000227.1 - 2 89
AK293847.1 - 10 1046
AK293879.1 - 15 1526
AK307914.1 - 4 481
BC015018.1 - 2 87
CR749492.1 - 19 1965
EU236710.2 - 44 4974

View Full Product Details