Gene Transcripts

Gene Symbol
Rhox3h
Entrez Gene ID
434758
Gene Name
reproductive homeobox 3H
Gene Aliases
EG434758, Rhox3.8, Rhox3g
Chromosome Location
Chr. X: 37657688 - 37668764 on Build GRCm38
UniGene ID
Mm.357753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114157.1 NP_001107629.1 6 939

View Full Product Details