Gene Transcripts

Gene Symbol
Ramp1
Entrez Gene ID
51801
Gene Name
receptor (calcitonin) activity modifying protein 1
Gene Aliases
9130218E19Rik
Chromosome Location
Chr. 1: 91179822 - 91225196 on Build GRCm38
UniGene ID
Mm.3272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168392.1 NP_001161864.1 2 430
NM_016894.3 NP_058590.1 2 430
NM_178401.3 NP_848488.2 2 430
GenBank mRNA AB019046.1 - 2 241
AF146522.1 - 2 222
AF209904.1 - 2 201
AJ250489.1 - 2 222
AK144331.1 - 2 430
AK157896.1 - 2 220
AK165238.1 - 2 206
BC012644.1 - 2 187 4207524
BY707433.1 - 2 211
BY733497.1 - 2 430
CB522711.1 - 1 67

View Full Product Details