Gene Transcripts

Gene Symbol
Rhox2h
Entrez Gene ID
622301
Gene Name
reproductive homeobox 2H
Gene Aliases
OTTMUSG00000017156, Rhox2.8, Rhox2g
Chromosome Location
Chr. X: 37668997 - 37673277 on Build GRCm38
UniGene ID
Mm.350896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100465.1 NP_001093935.1 3 530

Gene Symbol
Rhox2f
Entrez Gene ID
434764
Gene Name
reproductive homeobox 2F
Gene Aliases
EG434764, Rhox2.6, Rhox2e
Chromosome Location
Chr. X: 37571421 - 37576159 on Build GRCm38
UniGene ID
Mm.379498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085356.1 NP_001078825.1 3 530

View Full Product Details