Gene Transcripts

Gene Symbol
SEL1L3
Entrez Gene ID
23231
Gene Name
SEL1L family member 3
Gene Aliases
Sel-1L3
Chromosome Location
Chr. 4: 25697790 - 25863595 on Build GRCh38
UniGene ID
Hs.479384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297592.1 NP_001284521.1 20 2845
NM_001297594.1 NP_001284523.1 20 2867
NM_015187.4 NP_056002.2 20 3062
XM_011513819.2 XP_011512121.2 20 3056
GenBank mRNA AB018289.1 - 19 2560
AK025282.1 - 8 728
AK289687.1 - 20 2810
AK300219.1 - 11 1160
BC009945.2 - 13 1577
BC041046.1 - 11 1214
BC060867.1 - 20 2855 30344157
CR749817.1 - 20 2845

View Full Product Details