Gene Transcripts

Gene Symbol
Ccnb3
Entrez Gene ID
209091
Gene Name
cyclin B3
Gene Aliases
Chromosome Location
Chr. X: 6979652 - 7041635 on Build GRCm38
UniGene ID
Mm.448624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183015.3 NP_898836.2 7 1128
GenBank mRNA AJ555464.1 - 7 1129
BC132411.1 - 6 1082 40130688
BC138355.1 - 6 1082 8861375

View Full Product Details