Gene Transcripts
- Gene Symbol
- Ifnz
- Entrez Gene ID
- 319146
- Gene Name
- interferon zeta
- Gene Aliases
- 6030405N23Rik, BGIF, ENSMUSG00000073810, Lmtn
- Chromosome Location
- Chr. 4: 88782131 - 88783592 on Build GRCm38
- UniGene ID
- Mm.368232
- Species
- Mus musculus
- Gene Symbol
- Gm13272
- Entrez Gene ID
- 545648
- Gene Name
- predicted gene 13272
- Gene Aliases
- OTTMUSG00000011265
- Chromosome Location
- Chr. 4: 88779850 - 88780660 on Build GRCm38
- UniGene ID
- Mm.368246
- Species
- Mus musculus
- Gene Symbol
- Gm13276
- Entrez Gene ID
- 545649
- Gene Name
- predicted gene 13276
- Gene Aliases
- OTTMUSG00000011269
- Chromosome Location
- Chr. 4: 88785054 - 88786515 on Build GRCm38
- UniGene ID
- Mm.474424
- Species
- Mus musculus
- Gene Symbol
- Gm13277
- Entrez Gene ID
- 545650
- Gene Name
- predicted gene 13277
- Gene Aliases
- OTTMUSG00000011270
- Chromosome Location
- Chr. 4: 88787977 - 88789438 on Build GRCm38
- UniGene ID
- Mm.473677
- Species
- Mus musculus
- Gene Symbol
- Gm13278
- Entrez Gene ID
- 545651
- Gene Name
- predicted gene 13278
- Gene Aliases
- OTTMUSG00000011272
- Chromosome Location
- Chr. 4: 88790895 - 88792356 on Build GRCm38
- UniGene ID
- Mm.149203
- Species
- Mus musculus
- Gene Symbol
- Gm13279
- Entrez Gene ID
- 545653
- Gene Name
- predicted gene 13279
- Gene Aliases
- OTTMUSG00000011273
- Chromosome Location
- Chr. 4: 88797237 - 88798204 on Build GRCm38
- UniGene ID
- Mm.489586
- Species
- Mus musculus
- Gene Symbol
- Gm13283
- Entrez Gene ID
- 545645
- Gene Name
- predicted gene 13283
- Gene Aliases
- EG545645, OTTMUSG00000011280
- Chromosome Location
- Chr. 4: 88760774 - 88761584 on Build GRCm38
- UniGene ID
- Mm.489553
- Species
- Mus musculus
View Full Product Details