Gene Transcripts

Gene Symbol
Ncoa7
Entrez Gene ID
211329
Gene Name
nuclear receptor coactivator 7
Gene Aliases
9030406N13Rik
Chromosome Location
Chr. 10: 30645582 - 30805977 on Build GRCm38
UniGene ID
Mm.297903
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111267.2 NP_001104737.1 6 742
NM_172495.5 NP_766083.3 16 3093
GenBank mRNA AK033503.1 - 6 656
AK128935.1 - 15 2916
AK165098.1 - 4 1430
AK171679.1 - 5 586
BC042791.1 - 1173
BC049997.1 - 6 692
BC058572.1 - 6 664
BC076623.1 - 14 2699
BC080742.1 - 5 1848
BC141150.1 - 15 2794 9055762
BC145459.1 - 14 2679
EF602028.1 - 6 742

View Full Product Details