Gene Transcripts

Gene Symbol
Ncoa7
Entrez Gene ID
211329
Gene Name
nuclear receptor coactivator 7
Gene Aliases
9030406N13Rik
Chromosome Location
Chr. 10: 30645582 - 30805977 on Build GRCm38
UniGene ID
Mm.297903
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111267.2 NP_001104737.1 5 662
NM_172495.5 NP_766083.3 15 3013
GenBank mRNA AK033503.1 - 5 576
AK042369.1 - 5 632
AK128935.1 - 14 2836
AK165098.1 - 4 1035
BC042791.1 - 1093
BC049997.1 - 5 612
BC058572.1 - 5 584
BC076623.1 - 13 2619
BC080742.1 - 4 1768
BC141150.1 - 14 2714 9055762
BC145459.1 - 13 2599
EF602028.1 - 5 662

View Full Product Details