Gene Transcripts

Gene Symbol
Greb1l
Entrez Gene ID
381157
Gene Name
growth regulation by estrogen in breast cancer-like
Gene Aliases
mKIAA4095
Chromosome Location
Chr. 18: 10324963 - 10562941 on Build GRCm38
UniGene ID
Mm.261535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083628.1 NP_001077097.1 6 827
GenBank mRNA AK142556.1 - 6 830
BC147570.1 - 5 684 9056560

View Full Product Details